Benzo[b]thiophene-2-carbonitrile 95%

Supplier: Ambeed
Warning

A304752-10G A304752-1G A304752-250MG A304752-5G
76977-770EA 167 USD
76977-770 76977-772 76977-774 76977-776
Benzo[b]thiophene-2-carbonitrile 95%
Benzo[b]thiophene-2-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅NS
MDL Number: MFCD09751294
CAS Number: 55219-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR