4-(Fmoc-amino)-4-piperidinecarboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Fmoc-Amino)-4-piperidinecarboxylic acid hydrochloride

A106196-1G A106196-250MG A106196-5G
76970-448EA 54 USD
76970-448 76970-450 77570-360
4-(Fmoc-amino)-4-piperidinecarboxylic acid hydrochloride 95%
4-(Fmoc-amino)-4-piperidinecarboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₃ClN₂O₄
MW: 402.88 g/mol
MDL Number: MFCD03840417
CAS Number: 368866-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR