N-Boc-DL-valine 97%
Supplier: Ambeed
|
Warning
|
A179138-10G
A179138-25G
A179138-500G
A179138-5G
76974-378EA
9.55
USD
76974-378
76974-380
76974-382
76974-384
N-Boc-DL-valine 97%
N-Boc-DL-valine
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₄ MW: 217.26 g/mol |
MDL Number:
MFCD00069973 CAS Number: 54895-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...