tert-Butyl-3-(aminomethyl)benzoate 95%

Supplier: Ambeed
Danger

A554040-250MG A554040-1G
76980-244EA 24 USD
76980-244 77882-334
tert-Butyl-3-(aminomethyl)benzoate 95%
tert-Butyl-3-(aminomethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MDL Number: MFCD08275208
CAS Number: 102638-45-9
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR