6-Methoxy-2-methyl-3-nitropyridine 97%

Supplier: Ambeed
Warning

A196642-1G A196642-250MG A196642-25G A196642-5G
76975-080EA 7.43 USD
76975-080 76975-082 76975-084 76975-086
6-Methoxy-2-methyl-3-nitropyridine 97%
6-Methoxy-2-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.1500 g/mol
MDL Number: MFCD03095076
CAS Number: 5467-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR