1,1-Cyclobutanedicarboxylic acid 98%

Supplier: Ambeed
Danger

A262801-10G A262801-25G A262801-5G A262801-100G
76977-006EA 10 USD
76977-006 76977-008 76977-010 77881-715
1,1-Cyclobutanedicarboxylic acid 98%
1,1-Cyclobutanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₄
MDL Number: MFCD00001325
CAS Number: 5445-51-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR