Ethyl-2-bromooctanoate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-bromooctanoate

A715225-25G A715225-5G
76981-202EA 18.04 USD
76981-202 76981-204
Ethyl-2-bromooctanoate 98%
Ethyl-2-bromooctanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉BrO₂
MW: 251.164 g/mol
MDL Number: MFCD00042603
CAS Number: 5445-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR