2,6-Pyridinedicarboxaldehyde 98%

Supplier: Ambeed
Warning

A324883-100G A324883-100MG A324883-10G A324883-1G A324883-250MG A324883-25G A324883-5G
76978-112EA 979 USD
76978-112 76978-114 76978-116 76978-118 76978-120 76978-122 76978-124
2,6-Pyridinedicarboxaldehyde 98%
2,6-Pyridinedicarboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₅NO₂
MDL Number: MFCD00010103
CAS Number: 5431-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR