1-(Pyrazin-2-yl)ethanol 95%

Supplier: Ambeed
Warning

A778794-100MG A778794-250MG
76981-568EA 27 USD
76981-568 76981-570
1-(Pyrazin-2-yl)ethanol 95%
1-(Pyrazin-2-yl)ethanol

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD12153748
CAS Number: 94777-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR