3-Iodo-2-nitropyridine 98%

Supplier: Ambeed
Warning

A153219-1G A153219-250MG A153219-5G
76973-204EA 22 USD
76973-204 76973-206 76973-208
3-Iodo-2-nitropyridine 98%
3-Iodo-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃IN₂O₂
MDL Number: MFCD08061318
CAS Number: 54231-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR