tert-Butyl 3-(cyclopropylamino)pyrrolidine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A124577-100MG
A124577-250MG
A124577-1G
76971-774EA
63.66
USD
76971-774
76971-776
77670-766
tert-Butyl 3-(cyclopropylamino)pyrrolidine-1-carboxylate 98%
tert-Butyl 3-(cyclopropylamino)pyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₂N₂O₂ MW: 226.32 g/mol |
MDL Number:
MFCD06656604 CAS Number: 887587-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...