2-Chloro-N-ethylpyrimidin-4-amine 98%

Supplier: Ambeed
Warning

A383626-1G A383626-250MG
76978-850EA 91.24 USD
76978-850 76978-852
2-Chloro-N-ethylpyrimidin-4-amine 98%
2-Chloro-N-ethylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MW: 157.60 g/mol
MDL Number: MFCD09055369
CAS Number: 86443-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR