2-Bromo-4-(bromomethyl)pyridine 98%

Supplier: Ambeed
Danger

A582254-100MG A582254-250MG
76980-394EA 32.89 USD
76980-394 76980-396
2-Bromo-4-(bromomethyl)pyridine 98%
2-Bromo-4-(bromomethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
MDL Number: MFCD11847700
CAS Number: 83004-14-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR