L-Lysine-L-glutamate 95%

Supplier: Ambeed
Warning

Synonyms: H-Lys-OH H-Glu-OH salt, (S)-2,6-Diaminohexanoic acid (S)-2-aminopentanedioic acid salt

A100493-100G A100493-10G
76970-130EA 12 USD
76970-130 76970-132
L-Lysine-L-glutamate 95%
L-Lysine-L-glutamate

Precaución: for Research Use Only

Formula: C₁₁H₂₃N₃O₆
MDL Number: MFCD00038974
CAS Number: 5408-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR