3-Chloropropyldimethylammonium chloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Dimethylaminopropylchloride hydrochloride, 3-Dimethylaminopropyl chloride hydrochloride, N-(3-Chloropropyl)-N,N-dimethylammonium chloride, 3-Chloro-N,N-dimethylpropan-1-amine hydrochloride
A106129-1000G
A106129-100G
A106129-25G
A106129-500G
76970-440EA
131
USD
76970-440
76970-442
76970-444
76970-446
3-Chloropropyldimethylammonium chloride 97%
3-Chloropropyldimethylammonium chloride
Precaución: for Research Use Only
Formula:
C₅H₁₃Cl₂N |
MDL Number:
MFCD00012521 CAS Number: 5407-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...