3'-Bromo-2,2,2-trifluoroacetophenone 99%

Supplier: Ambeed
Warning

A976670-100MG A976670-1G A976670-250MG A976670-25G A976670-5G
76982-048EA 5 USD
76982-048 76982-050 76982-052 76982-054 77614-696
3'-Bromo-2,2,2-trifluoroacetophenone 99%
3'-Bromo-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
MDL Number: MFCD00511232
CAS Number: 655-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR