2-Chloro-5-fluorobenzamide 97%

Supplier: Ambeed
Warning

A538918-1G A538918-250MG A538918-5G
76980-156EA 75.33 USD
76980-156 76980-158 76980-160
2-Chloro-5-fluorobenzamide 97%
2-Chloro-5-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅ClFNO
MW: 173.57 g/mol
MDL Number: MFCD01631465
CAS Number: 88487-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR