N1,N2-Dimethylcyclohexane-1,2-diamine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A327540-1G
A327540-250MG
A327540-25G
A327540-5G
76978-192EA
24.41
USD
76978-192
76978-194
76978-196
76978-198
N1,N2-Dimethylcyclohexane-1,2-diamine hydrochloride 95%
N1,N2-Dimethylcyclohexane-1,2-diamine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₈N₂·ClH MW: 178.703 g/mol |
MDL Number:
MFCD21603997 CAS Number: 1956325-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...