Ethyl-3-amino-2,6-dichloroisonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-amino-2,6-dichloropyridine-4-carboxylate, Ethyl 3-amino-2,6-dichloroisonicotinate

A501705-1G A501705-250MG A501705-5G
76979-866EA 97.61 USD
76979-866 76979-868 76979-870
Ethyl-3-amino-2,6-dichloroisonicotinate 97%
Ethyl-3-amino-2,6-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₈H₈Cl₂N₂O₂
MW: 235.07 g/mol
MDL Number: MFCD16661134
CAS Number: 458543-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR