(2-Chlorophenyl)acetamide 98%

Supplier: Ambeed
Warning

A964684-1G A964684-5G
76982-028EA 34 USD
76982-028 76982-030
(2-Chlorophenyl)acetamide 98%
(2-Chlorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MDL Number: MFCD00176724
CAS Number: 10268-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR