Benzyl (2,2-dimethoxyethyl)carbamate 96%

Supplier: Ambeed
Warning

A601108-100MG A601108-1G A601108-250MG
76980-508EA 5 USD
76980-508 76980-510 76980-512
Benzyl (2,2-dimethoxyethyl)carbamate 96%
Benzyl (2,2-dimethoxyethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₄
MW: 239.26 g/mol
MDL Number: MFCD13184754
CAS Number: 114790-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR