Tetrahydropyran-4-sulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: Tetrahydro-2H-pyran-4-sulphonyl chloride, 2H-Pyran-4-sulphonylchloride,tetrahydro-, oxane-4-sulphonyl chloride

A897868-250MG A897868-1G
76981-904EA 47 USD
76981-904 77573-804
Tetrahydropyran-4-sulfonyl chloride 95%
Tetrahydropyran-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₅H₉ClO₃S
MDL Number: MFCD07787616
CAS Number: 338453-21-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR