Ethyl-2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic Acid Ethyl Ester, Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 2-Amino-3-carbethoxy-4,5-tetramethylenethiophene, 2-Amino-3-ethoxycarbonyl-4,5-tetramethylenethiophene

A148097-100G A148097-25G A148097-1G A148097-5G
76972-932EA 91 USD
76972-932 76972-934 77881-153 77881-154
Ethyl-2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate 98%
Ethyl-2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂S
MDL Number: MFCD00005862
CAS Number: 4506-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR