4-(5-Bromo-1,3,4-thiadiazol-2-yl)morpholine 95%

Supplier: Ambeed
Warning

A309153-250MG A309153-5G
76977-862EA 67.9 USD
76977-862 76977-864
4-(5-Bromo-1,3,4-thiadiazol-2-yl)morpholine 95%
4-(5-Bromo-1,3,4-thiadiazol-2-yl)morpholine

Precaución: for Research Use Only

Formula: C₆H₈BrN₃OS
MW: 250.1200 g/mol
MDL Number: MFCD12923103
CAS Number: 1341405-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR