3-Ethoxy-4-fluorobenzaldehyde 96%

Supplier: Ambeed
Warning

A944012-100MG A944012-250MG A944012-5G
76981-996EA 40.32 USD
76981-996 76981-998 76982-000
3-Ethoxy-4-fluorobenzaldehyde 96%
3-Ethoxy-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₂
MW: 168.165 g/mol
MDL Number: MFCD12022599
CAS Number: 870837-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR