Cinacalcet hydrochloride 98%

Supplier: Ambeed

Synonyms: (R)-N-(1-(Naphthalen-1-yl)ethyl)-3-(3-(trifluoromethyl)phenyl)propan-1-amine hydrochloride

A164114-100MG A164114-10MG A164114-1G A164114-250MG A164114-50MG A164114-5G A164114-5MG A164114-25G A164114-100G
76973-700EA 14 USD
76973-700 76973-702 76973-704 76973-706 76973-708 76973-710 76973-712 77671-312 77881-274
Cinacalcet hydrochloride 98%
Cinacalcet hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₂F₃N·HCl
MW: 393.88 g/mol
MDL Number: MFCD08067750
CAS Number: 364782-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR