Ethyl-2-amino-5-methoxybenzoate 97%

Supplier: Ambeed
Warning

A798107-1G A798107-250MG A798107-5G
76981-652EA 43 USD
76981-652 76981-654 76981-656
Ethyl-2-amino-5-methoxybenzoate 97%
Ethyl-2-amino-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD11111632
CAS Number: 64018-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR