cis-4-Hydroxycyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

A713991-100G A713991-10G A713991-1G A713991-250MG A713991-25G A713991-5G
76981-180EA 1330.37 USD
76981-180 76981-182 76981-184 76981-186 76981-188 76981-190
cis-4-Hydroxycyclohexanecarboxylic acid 98%
cis-4-Hydroxycyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MW: 144.1700 g/mol
MDL Number: MFCD02093477
CAS Number: 3685-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR