6-Bromo-1H-pyrazolo[4,3-b]pyridin-3-amine 97%
Supplier: Ambeed
|
Warning
|
A201146-100MG
A201146-1G
A201146-250MG
A201146-5G
76975-264EA
29.71
USD
76975-264
76975-266
76975-268
76975-270
6-Bromo-1H-pyrazolo[4,3-b]pyridin-3-amine 97%
6-Bromo-1H-pyrazolo[4,3-b]pyridin-3-amine
Precaución: for Research Use Only
Formula:
C₆H₅BrN₄ |
MDL Number:
MFCD08062675 CAS Number: 1211516-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...