2-Phenylisoindolin-1-one 97%

Supplier: Ambeed
Warning

A801405-1G A801405-250MG
76981-662EA 167 USD
76981-662 76981-664
2-Phenylisoindolin-1-one 97%
2-Phenylisoindolin-1-one

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
MDL Number: MFCD00023020
CAS Number: 5388-42-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR