6-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

A912781-1G A912781-5G
76981-922EA 40.32 USD
76981-922 76981-924
6-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one 95%
6-Bromo-4-(trifluoromethyl)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃NO
MDL Number: MFCD11846359
CAS Number: 328955-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR