4-(Dimethylamino)benzamide 97%

Supplier: Ambeed
Warning

A152920-1G A152920-250MG A152920-100MG
76973-172EA 58 USD
76973-172 76973-174 77881-178
4-(Dimethylamino)benzamide 97%
4-(Dimethylamino)benzamide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O
MDL Number: MFCD01216782
CAS Number: 6083-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR