4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 96%

Supplier: Ambeed
Warning

A319795-1G A319795-250MG A319795-5G
76978-042EA 44.56 USD
76978-042 76978-044 76978-046
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 96%
4-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClN₃
MW: 177.59 g/mol
MDL Number: MFCD11046237
CAS Number: 920965-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR