4,6-Difluorosalicylaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2,4-Difluoro-6-hydroxybenzaldehyde

A129156-250MG A129156-5G A129156-1G A129156-100MG
76971-972EA 17 USD
76971-972 77611-144 77670-828 77881-030
4,6-Difluorosalicylaldehyde 97%
4,6-Difluorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
MDL Number: MFCD03701048
CAS Number: 136516-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR