(R)-3-Amino-1-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butan-1-one phosphate 98%
Supplier: Ambeed
|
Warning
|
A111654-1G
A111654-5G
A111654-100MG
A111654-250MG
76970-838EA
21
USD
76970-838
76970-840
77610-882
77610-884
(R)-3-Amino-1-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butan-1-one phosphate 98%
(R)-3-Amino-1-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butan-1-one phosphate
Precaución: for Research Use Only
Formula:
C₁₆H₁₈F₆N₅O₅P |
MDL Number:
MFCD09952339 CAS Number: 654671-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...