3,4-Dehydro-L-proline 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2,5-Dihydro-1H-pyrrole-2-carboxylic acid, (S)-3-Pyrroline-2-carboxylic Acid, H-3,4-Dehydro-Pro-OH

A170862-100MG A170862-1G A170862-250MG
76974-006EA 40 USD
76974-006 76974-008 76974-010
3,4-Dehydro-L-proline 95%
3,4-Dehydro-L-proline

Precaución: for Research Use Only

Formula: C₅H₇NO₂
MW: 113.12 g/mol
MDL Number: MFCD00065964
CAS Number: 4043-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR