Methyl 3-bromo-6-chloropyrazine-2-carboxylate 95%

Supplier: Ambeed
Warning

A181274-1G A181274-250MG
76974-470EA 152.77 USD
76974-470 76974-472
Methyl 3-bromo-6-chloropyrazine-2-carboxylate 95%
Methyl 3-bromo-6-chloropyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄BrClN₂O₂
MW: 251.46 g/mol
MDL Number: MFCD13195362
CAS Number: 13457-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR