3,5-Difluoro-4-hydroxybenzonitrile 96%

Supplier: Ambeed
Danger

A471678-1G A471678-250MG A471678-5G
76979-676EA 49.87 USD
76979-676 76979-678 77613-458
3,5-Difluoro-4-hydroxybenzonitrile 96%
3,5-Difluoro-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃F₂NO
MW: 155.1 g/mol
MDL Number: MFCD09033181
CAS Number: 2967-54-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR