2-Bromo-3-methylbenzotrifluoride 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-methyl-3-(trifluoromethyl)benzene

A116208-1G A116208-250MG A116208-5G
76971-192EA 155.96 USD
76971-192 76971-194 76971-196
2-Bromo-3-methylbenzotrifluoride 95%
2-Bromo-3-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD11100539
CAS Number: 1099597-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR