4-(1-Naphthyl)-1-naphthaleneboronic acid 98%

Supplier: Ambeed
Warning

A284584-250MG A284584-1G A284584-5G
76977-490EA 17 USD
76977-490 77881-778 77881-779
4-(1-Naphthyl)-1-naphthaleneboronic acid 98%
4-(1-Naphthyl)-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₅BO₂
MDL Number: MFCD16294426
CAS Number: 363607-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR