[1,1'-Biphenyl]-2,2'-diyldiboronic acid 95%

Supplier: Ambeed
Warning

A513921-100MG A513921-1G A513921-250MG
76979-938EA 95 USD
76979-938 76979-940 76979-942
[1,1'-Biphenyl]-2,2'-diyldiboronic acid 95%
[1,1'-Biphenyl]-2,2'-diyldiboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂B₂O₄
MDL Number: MFCD21603686
CAS Number: 312968-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR