4,4,5,5-Tetramethyl-2-(4-(naphthalen-2-yl)phenyl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A243722-250MG A243722-25G A243722-5G
76976-536EA 6 USD
76976-536 76976-538 76976-540
4,4,5,5-Tetramethyl-2-(4-(naphthalen-2-yl)phenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(4-(naphthalen-2-yl)phenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₂H₂₃BO₂
MDL Number: MFCD09800562
CAS Number: 1092390-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR