9,9'-Spirobi[9H-fluorene]-2-boronic acid 95%

Supplier: Ambeed
Warning

A194023-1G A194023-250MG
76974-960EA 13 USD
76974-960 76974-962
9,9'-Spirobi[9H-fluorene]-2-boronic acid 95%
9,9'-Spirobi[9H-fluorene]-2-boronic acid

Precaución: for Research Use Only

Formula: C₂₅H₁₇BO₂
MDL Number: MFCD13248568
CAS Number: 236389-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR