L-Tyrosine benzyl ester-p-toluenesulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-Tyrosine benzyl ester p-toluenesulphonate salt, H-Tyr-OBzl·TosOH, H-Tyr-OBzl.Tos-OH
A456652-25G
A456652-100G
A456652-5G
A456652-500G
A456652-1G
76979-522EA
17
USD
76979-522
77613-410
77673-148
77673-146
77882-170
L-Tyrosine benzyl ester-p-toluenesulfonate 98%
L-Tyrosine benzyl ester-p-toluenesulfonate
Precaución: for Research Use Only
Formula:
C₁₆H₁₇NO₃ |
MDL Number:
MFCD00038959 CAS Number: 53587-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...