3-Amino-4-nitrobenzonitrile 95%

Supplier: Ambeed
Danger

A859211-1G A859211-250MG A859211-5G
76981-828EA 50 USD
76981-828 76981-830 76981-832
3-Amino-4-nitrobenzonitrile 95%
3-Amino-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MDL Number: MFCD18389919
CAS Number: 99512-10-4
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR