2-Nitro-4-(trifluoromethyl)benzaldehyde 98%

Supplier: Ambeed
Warning

A998570-1G A998570-5G A998570-10G A998570-250MG
76982-098EA 28 USD
76982-098 76982-100 77882-867 77882-868
2-Nitro-4-(trifluoromethyl)benzaldehyde 98%
2-Nitro-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₃
MDL Number: MFCD06797985
CAS Number: 109466-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR