Amino-PEG4-acid 95%

Supplier: Ambeed
Warning

Synonyms: Amino-PEG4-acid, Amino-PEG-acid, α-Amine-ω-propionic acid tetraethylene glycol, 3-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]propionic acid, NH2-PEG4-COOH, Carboxy-PEG4-amine, Tetraethylene glycol amino propionic acid, NH2-PEG4-CH2CH2COOH, CA(PEG)4, (PEO)4-Aminopropionic acid, 1-Amino-3,6,9,12-tetraoxapentadecan-15-oic acid, Tetraethylene glycol α-amino-ω-propionic acid

A125981-100MG A125981-1G A125981-250MG A125981-5G
76971-874EA 14 USD
76971-874 76971-876 76971-878 76971-880
Amino-PEG4-acid 95%
Amino-PEG4-acid

Precaución: for Research Use Only

Formula: C₁₁H₂₃NO₆
MDL Number: MFCD11041129
CAS Number: 663921-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR