2-Bromo-4-methoxypyridin-3-amine 97%

Supplier: Ambeed
Warning

A176080-100MG A176080-250MG A176080-5G A176080-1G
76974-254EA 77.45 USD
76974-254 76974-256 76974-258 77593-120
2-Bromo-4-methoxypyridin-3-amine 97%
2-Bromo-4-methoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MW: 203.04 g/mol
MDL Number: MFCD11044243
CAS Number: 109613-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR