3-Hydroxy-2-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A319922-100MG A319922-1G A319922-250MG
76978-048EA 56.23 USD
76978-048 76978-050 76978-052
3-Hydroxy-2-nitrobenzonitrile 95%
3-Hydroxy-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
MW: 164.12 g/mol
MDL Number: MFCD09834347
CAS Number: 129298-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR