3-(Methylsulfonyl)benzoic acid 97%

Supplier: Ambeed
Warning

A131518-100G A131518-10G A131518-1G A131518-250MG A131518-5G
76972-102EA 320.4 USD
76972-102 76972-104 76972-106 76972-108 76972-110
3-(Methylsulfonyl)benzoic acid 97%
3-(Methylsulphonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₄S
MW: 200.21 g/mol
MDL Number: MFCD00216488
CAS Number: 5345-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR